missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NI 57
GREENER_CHOICE

Product Code. p-7110843
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700518 10 mg 10mg
18744059 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18700518 Supplier Tocris Bioscience™ Supplier No. 5546/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective BRPF bromodomain inhibitor

NI 57 is a potent and selective BRPF bromodomain inhibitor (Kd values are 31, 108 and 408 nM, for BRPF1B, 2 and 3, respectively, which exhibits >32-fold selectivity for BRPFs over BRD9 and other non-Class IV bromodomains. NI 57 inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Cyano-N-(1,3-dimethyl-2-oxo-1,2-dihydroquinolin-6-yl)-2-methoxybenzene-1-sulfonamide
CAS 1883548-89-7
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C19H17N3O4S
Formula Weight 383.42
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.