missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nick Translation System

Product Code. 10634623
Change view
Click to view available options
Quantity:
50 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10634623 50 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10634623 Supplier Invitrogen™ Supplier No. 18160010
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nick Translation System

The Nick Translation System is ideal for both radioactive and non-radioactive labeling of DNA. The Nick Translation System:

  • Provides five pre-mixed nucleotide solutions for flexibility in labeled nucleotide incorporation
  • Yields >108 cpm/μg control DNA using [ - 32 P]-dCTP
  • Labels 1μg of DNA in one reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The Nick Translation System includes an enzyme mix containing DNA Polymerase I and DNase I, dNTPs, control DNA, stop buffer, and distilled water. Store at -20°C. Guaranteed stable for 6 months when properly stored.
    Detection Method Radioactive, Fluorescence
    Final Product Type Probes (Labeled DNA)
    Labeling Target DNA (General)
    Includes Label or Dye No
    Labeling Method Direct Labeling
    Product Type Nick Translation System
    Quantity 50 Reactions
    Shipping Condition Wet Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.