missing translation for 'onlineSavingsMsg'
Learn More

Nickel 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 11384498
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384498 25 mL 25mL
11394498 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384498 Supplier Thermo Scientific Alfa Aesar Supplier No. 042378.AC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142600-62-2
Molecular Formula C6H14NiO4
Molecular Weight (g/mol) 208.867
MDL Number MFCD01863472
InChI Key KNEAYFOSQJIPKZ-UHFFFAOYSA-N
Synonym nickel 2+ bis 2-methoxyethanolate
PubChem CID 131873637
IUPAC Name 2-methoxyethanolate;nickel(2+)
SMILES COCC[O-].COCC[O-].[Ni+2]

Specifications

Packaging ChemSeal™ bottles
Linear Formula Ni(OCH2CH2OCH3)2
Quantity 25 mL
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 208.76
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Chemical Name or Material Nickel 2-methoxyethoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.