missing translation for 'onlineSavingsMsg'
Learn More

Nickel acetylacetonate, 96%

Product Code. 10437710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10437710 25 g Glass bottle 25g
10251123 5 g Glass bottle 5g
10371094 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437710 Supplier Thermo Scientific Acros Supplier No. 128260250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3264-82-2 | C10H14NiO4 | 256.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
IUPAC Name nickel(2+);(E)-4-oxopent-2-en-2-olate
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 230°C
Color Yellow
Boiling Point 220°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Complexometry)
Packaging Glass bottle
Quantity 25 g
Fieser 05,471; 06,417; 07,250; 08,42; 17,201
Merck Index 15, 6585
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin
Formula Weight 256.9
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Nickel acetylacetonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.