missing translation for 'onlineSavingsMsg'
Learn More

Nickel aluminum, 99.0% min (metals basis)

Product Code. 11338117
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338117 50 g 50g
11328117 10 g 10g
11318117 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11338117 Supplier Thermo Scientific Alfa Aesar Supplier No. 022890.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 12003-78-0 | AlNi | 85.675 g/mol

Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-78-0
Molecular Formula AlNi
Molecular Weight (g/mol) 85.675
MDL Number MFCD00167392
InChI Key NPXOKRUENSOPAO-UHFFFAOYSA-N
Synonym aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel
PubChem CID 6336846
IUPAC Name aluminum;nickel
SMILES [Al].[Ni]

Specifications

Color Gray
Appearance Gray powder, Intermetallic
Quantity 50 g
Odor Odorless
Assay Percent Range 99.0% min. (metals basis)
Form Powder
Solubility Information Insoluble in water.
Chemical Name or Material Nickel aluminum
compliance-icons compliance-icons
Product Identifier
  • Nickel aluminum
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Aluminum, compound with nickel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.