missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals

Product Code. 11495021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495021 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495021 Supplier MP Biomedicals Supplier No. 0215582591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell Culture Reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2 · 6H2O

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 237.7
Chemical Name or Material Nickel Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.