missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

Product Code. 15474805
Change view
Click to view available options
Quantity:
1 Ea.
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity missing translation for 'unitSize'
15474805 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15474805 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 040870.KS
 Restricted Item
Estimated Shipment: 18-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 11106-97-1 | CrNi | 110.69 g/mol

Used as Magnetron sputtering source. The substrate bias during dc sputtering effectively reduces oxygen contamination in nichrome films. This in turn leads to an enhancement of the preferred orientation of the deposits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Quantity 1 Ea.
CAS 11106-97-1
Odor Odorless
Molecular Formula CrNi
MDL Number MFCD00673110
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
Solubility Information Insoluble in water.
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES [Cr].[Ni]
IUPAC Name chromium;nickel
Molecular Weight (g/mol) 110.69
PubChem CID 11320920
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Nickel Chromium, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.