missing translation for 'onlineSavingsMsg'
Learn More

Nickel dimethylglyoxime, 99%, Thermo Scientific™

Product Code. 11338258
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338258 2 g 2g
11358258 10 g 10g
11348258 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338258 Supplier Thermo Scientific Alfa Aesar Supplier No. 039458.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Paint pigment

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-93-8
Molecular Formula C8H14N4NiO4
Molecular Weight (g/mol) 288.92
MDL Number MFCD00136028
InChI Key UNMGLSGVXHBBPH-USBCBBGOSA-L
Synonym nickel dimethylglyoxime,, nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine
PubChem CID 15639912
SMILES [Ni++].C\C(=N\O)\C(\C)=N/[O-].C\C(=N\O)\C(\C)=N/[O-]

Specifications

Melting Point 250°C (sublimation)
Quantity 2 g
Solubility Information Insoluble in water; Soluble in mineral acids
IUPAC Name nickel(2+) bis((Z)-[(3Z)-3-(hydroxyimino)butan-2-ylidene](oxido)amine)
Formula Weight 288.94
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Nickel dimethylglyoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.