missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 1.0mm (0.04in) thick, Puratronic™, 99.9945% (metals basis)

Product Code. 11336388
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
Product Code. Quantity unitSize
11336388 100 x 100 mm Each
11326388 25 x 25 mm 25mm
11375899 50 x 50 mm Each
3 options
This item is not returnable. View return policy

Product Code. 11336388

Brand: Thermo Scientific Alfa Aesar 012043.GH

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈5.6g/25x25mm
Form Foil
Percent Purity 99.9945%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Nickel foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.