missing translation for 'onlineSavingsMsg'
Learn More

Nickel hydroxyacetate, Ni 27-29%, Thermo Scientific™

Product Code. 11328258
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328258 100 g 100g
11318258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328258 Supplier Thermo Scientific Alfa Aesar Supplier No. 039456.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41587-84-2
Molecular Formula C4H6NiO6
Molecular Weight (g/mol) 208.779
MDL Number MFCD00070414
InChI Key OZHLXQFAHIYYDJ-UHFFFAOYSA-L
Synonym nickel hydroxyacetate, acmc-20ak3m, nickel 2+ diglycolate, nickel 2+ bis hydroxyacetate, nickel 2+ ion diglycolate, bis hydroxyacetic acid nickel ii salt, nickel, 2,2'-bipyridine-n,n' hydroxyacetato 2--o1,o2-9ci
PubChem CID 57349863
IUPAC Name 2-hydroxyacetate;nickel(2+)
SMILES C(C(=O)[O-])O.C(C(=O)[O-])O.[Ni+2]

Specifications

Color Green
Linear Formula Ni(OOCCH2OH)2
Quantity 100 g
UN Number UN3077
Formula Weight 208.78
Concentration or Composition (by Analyte or Components) Ni 27 to 29%
Physical Form Powder
Chemical Name or Material Nickel hydroxyacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.