missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) 2,4-pentanedionate, 95%

Product Code. 11323487
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323487 25 g 25g
11393477 5 g 5g
11303487 100 g 100g
11313487 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11323487 Supplier Thermo Scientific Alfa Aesar Supplier No. 018811.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Catalyst and U.V. stabilizer | CAS: 3264-82-2 | C10H14NiO4 | 256.91 g/mol

Catalyst and U.V. stabilizer.Nickel(II) 2,4-pentanedionate is used as a precursor to nickel bis(cyclooctadiene) catalyst. It is also used in the deposition of nickel(II) oxide thin film by sol-gel techniques on conductive glass substrates. Further, it is used in organic synthesis to produce organometals. It is associated with dimethylgold(III) acetylacetonate is used in gold on nickel plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼230°C (decomposition)
Color Green
Odor Odorless
Quantity 25 g
Beilstein 4157970
Sensitivity Hygroscopic
Merck Index 14,6500
Solubility Information Soluble in water,alcohol,toluene.
IUPAC Name nickel(2+);(E)-4-oxopent-2-en-2-olate
Formula Weight 256.91
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nickel(II) 2,4-pentanedionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.