Learn More
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
Description
It is used in electroplating, anodize coatings; textile dyeing mordant; Catalyst. Also, used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | Green |
| Physical Form | Crystalline Aggregates |
| Quantity | 5 g |
| Linear Formula | Ni(CH3COO)2·4H2O |
| UN Number | UN3077 |
| Beilstein | 3730764 |
| Merck Index | 14,6499 |
| Solubility Information | Soluble in water and methanol. Insoluble in diethyl ether,n-octanol. |
| Formula Weight | 248.84 (176.79 Anhydrous) |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.