missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Acetate Tetrahydrate, TRC

Product Code. 30469140
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30469140 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30469140

Brand: Toronto Research Chemicals TRCN394570100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel(II) Acetate Tetrahydrate
CAS 6018-89-9
Molecular Formula C4H16NiO8
Quantity 100 g
Synonym Acetic Acid Nickel(2+) Salt Tetrahydrate, Diacetatonickel TetrahydrateNickel Acetate [Ni(OAc)2.4H2O] Tetrahydrate, Nickel Acetate Tetrahydrate, Nickel Diacetate Tetrahydrate, Nickel(2+) Acetate Tetrahydrate, Nickel(2+) Diacetate Tetrahydrate
InChI Formula InChI=1S/2C2H4O2.Ni.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2
SMILES O.O.O.O.[Ni+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name nickel(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 250.86
Formula Weight 248.0042
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.