missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Bromide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Nickel(II) Bromide |
| CAS | 13462-88-9 |
| Molecular Formula | Br2Ni |
| Quantity | 25 g |
| Synonym | Nickel dibromide, Nickel(2+) bromide, Nickel bromideNSC 128153, Nickel bromide (Br2Ni) |
| InChI Formula | InChI=1S/2BrH.Ni/h2*1H;/q;;+2/p-2 |
| SMILES | Br[Ni]Br |
| IUPAC Name | dibromonickel |
| Molecular Weight (g/mol) | 218.5 |
| Formula Weight | 215.772 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction