Learn More
Nickel(II) bromide trihydrate, 98%
Description
It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 300°C (in -3H2O) |
| Physical Form | Crystalline Granules |
| Quantity | 250 g |
| UN Number | UN3077 |
| Merck Index | 14,6502 |
| Solubility Information | Soluble in water, alcohol |
| Formula Weight | 272.58 (218.53 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.