missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Anhydrous, TRC

Product Code. 30477613
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477613 500 mg -
30479225 250 mg 250mg
30470036 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477613 Supplier Toronto Research Chemicals Supplier No. TRCN395053500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula NiCl2
Molecular Weight (g/mol) 129.6
IUPAC Name nickel(2+);dichloride
SMILES [Cl-].[Cl-].[Ni+2]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2ClH.Ni/h2*1H;/q;;+2/p-2
Formula Weight 127.8731
Chemical Name or Material Nickel(II) Chloride Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.