missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 98+%

Product Code. 10534284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10534284 250 g Glass bottle 250g
10741141 1 kg Glass bottle 1kg
11934181 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10534284 Supplier Thermo Scientific Acros Supplier No. 270512500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni·6H2O
Molecular Weight (g/mol) 237.71

Specifications

Color Green
pH 4 to 6 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula NiCl2·6H2O
Fieser 14,351; 15,96; 17,86
Merck Index 15,659
Solubility Information Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Formula Weight 237.71
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Nickel(II) chloride hexahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Nickel(II) chloridehexahydrate (1:2:6),Nickel(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.