missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 98+%

Product Code. 11934181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
250g
5kg
Product Code. Quantity Packaging unitSize
11934181 5 kg Plastic Bottle 5kg
10534284 250 g Glass bottle 250g
10741141 1 kg Glass bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 11934181

Brand: Thermo Scientific Acros 270510050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni·6H2O
Molecular Weight (g/mol) 237.71

Specifications

Color Green
pH 4 to 6 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 98% min. (Complexometry)
Linear Formula NiCl2·6H2O
Fieser 14,351; 15,96; 17,86
Merck Index 15,659
Solubility Information Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Formula Weight 237.71
Percent Purity ≥98%
Packaging Plastic Bottle
Chemical Name or Material Nickel(II) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.