missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Hexahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10316290 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10316290 1 kg 1kg
10080260 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316290 Supplier Fisher Chemical Supplier No. N/1650/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni·6H2O
MDL Number 149809

Specifications

Color Green
pH 4 to 6
Physical Form Solid
Quantity 1 kg
Formula Weight 237.71g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 615.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.