missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Hexahydrate, puriss. p.a., ≥98%, Solstice

Product Code. 15633460
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15633460 2.5 kg Poly Bottle 2.5kg
15623460 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15633460 Supplier Honeywell-Fluka Supplier No. 314622.5KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Puriss. p.a.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-20-0
MDL Number MFCD00149809

Specifications

Melting Point 80°C
Quantity 2.5 kg
Linear Formula NiCl2 · 6H2O
UN Number UN3288
Formula Weight 237.69g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Packaging Poly Bottle
Density 3.550g/cm3
Chemical Name or Material Nickel(Ii) Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.