Learn More
Nickel(II) fluoride, anhydrous, 97%
Description
Nickel fluoride comprises the passivating surface that forms on nickel alloys, e.g. monel, which is why such materials are good to store or transport hydrogen fluoride or elemental fluorine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Crystalline |
| Quantity | 50 g |
| UN Number | UN3288 |
| Merck Index | 14,6508 |
| Solubility Information | Slightly soluble in water |
| Formula Weight | 96.71 |
| Percent Purity | 97% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.