missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) fluoride tetrahydrate, 98+%

Product Code. 11347737
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11347737

Brand: Thermo Scientific Alfa Aesar 014132.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13940-83-5

Nickel(II) fluoride tetrahydrate is used in surface treatment of aluminum profile and printing ink making, fluorescence lamp, fluidizer and catalyzer for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13940-83-5
MDL Number MFCD00016263

Specifications

Physical Form Powder
Quantity 250 g
UN Number UN3288
Solubility Information Soluble in water,inorganic acid.
Formula Weight 168.77 (96.71 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Chemical Name or Material Nickel(II) fluoride tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.