Learn More
Nickel(II) octanoate, in mineral spirits (8% Ni)
Description
Nickel(II) octanoate is used to prepare nickel based catalysts using niobia modified silica as support. It is also used as a catalyst and involved in the treatment of qaiyarah heavy crude oil, petrolene using ethanol as a solvent and reductant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Density | 0.95 g/mL |
| Flash Point | 40°C (104°F) |
| Linear Formula | [CH3(CH2)6COO]2Ni |
| Quantity | 500 g |
| UN Number | UN1993 |
| Solubility Information | Miscible with ethanol,chloroform,acetonitrile,ether,carbon disulfide,petroleum ether and glacial acetic acid. Immiscible with water. |
| Formula Weight | 345.1 |
| Concentration or Composition (by Analyte or Components) | 8% Ni |
| Physical Form | Liquid |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.