missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, 97%, green, reagent, powder

Product Code. 10126763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
10126763 100 g Glass bottle 100g
10740292 500 g Glass bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 10126763

Brand: Thermo Scientific Acros 415581000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1960°C
Color Green
CAS Min % 96.0
CAS Max % 100.0
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min. (Complexometry)
Linear Formula NiO
Merck Index 15, 6597
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in caustic solutions,soluble in acids and ammonium hydroxide
Formula Weight 74.7
Percent Purity 97%
Packaging Glass bottle
Specific Gravity 6.67
Density 6.6700g/mL
Chemical Name or Material Nickel(II) oxide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) oxide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H350i-May cause cancer by inhalation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.