missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, black, 76% Ni

Product Code. 10471261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
Product Code. Quantity Packaging unitSize
10471261 500 g Glass bottle 500g
10501091 5 g Glass bottle 5g
10318550 100 g Glass bottle 100g
10300744 2.5 kg Plastic bottle 2.5kg
4 options
This item is not returnable. View return policy

Product Code. 10471261

Brand: Thermo Scientific Acros 223125000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Color Black-Gray
Physical Form Powder
Quantity 500 g
Linear Formula NiO
Merck Index 15, 6597
Formula Weight 74.7
Percent Purity 75 to 77% (Ni)
Packaging Glass bottle
Density 6.67g/mL
Chemical Name or Material Nickel(II) oxide, black, 76% Ni

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.