missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3})

Product Code. p-7080367
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11324969

Brand: Thermo Scientific Alfa Aesar 087302.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

Nickel(II) oxide is used in fuel cell electrodes, storage batteries, electronic circuit boards, electrical ceramics, and as a pigment for ceramics and glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Color Black
Physical Form Powder
Quantity 250 g
Merck Index 14,6512
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 74.71
Concentration or Composition (by Analyte or Components) Ni 76%, (contains Ni2O3)
Odor Odorless
Density 6.67 g/mL
Chemical Name or Material Nickel (II) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.