missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, Puratronic™, 99.998% (metals basis)

Product Code. 11336947
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11336947

missing translation for 'mfr': Thermo Scientific Alfa Aesar 010819.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-99-1 | NiO | 74.69 g/mol

Nickel (II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in ammonium hydroxide. Insoluble in water.
Formula Weight 74.71
Density 6.67
Chemical Name or Material Nickel(II) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.