missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade

Product Code. 11450281
Change view
Click to view available options
Quantity:
1 L
2 x 2.5 L
Unit Size:
1 L
5 L
Product Code. Quantity unitSize
11450281 2 x 2.5 L 5 L
11308488 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 11450281

Brand: Thermo Scientific Alfa Aesar 040501.8M

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel plating | CAS: 13770-89-3 | H4N2NiO6S2 | 250.85 g/mol

Nickel plating Nickel(II) sulfamate is used for electroplating and as a surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-89-3
Molecular Formula H4N2NiO6S2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00137261
InChI Key KERTUBUCQCSNJU-UHFFFAOYSA-L
Synonym nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
PubChem CID 83720
IUPAC Name nickel(2+);disulfamate
SMILES [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

Color Green
Physical Form Liquid
Quantity 2 x 2.5 L
Linear Formula Ni(SO3NH2)2
UN Number UN3082
Solubility Information Miscible with water,immiscible with ether,acetone.
Formula Weight 250.86 (Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Nickel(II) sulfamate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.