missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)

Product Code. 11346947
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346947 25 g 25g
11356947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346947 Supplier Thermo Scientific Alfa Aesar Supplier No. 010820.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15244-37-8 | NiO4S | 154.749 g/mol

Nickel(II) sulfate hydrate is used in electroplating and as a chemical intermediate to produce other nickel compounds. It is also used in printing and in dyeing of textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-37-8
Molecular Formula NiO4S
Molecular Weight (g/mol) 154.749
MDL Number MFCD00011146
InChI Key LGQLOGILCSXPEA-UHFFFAOYSA-L
Synonym nickel sulfate, nickel sulphate, nickel ii sulfate, nickelous sulfate, nickel 2+ sulfate, nickel sulfate 1:1, niso4, nickel ii sulfate 1:1, nickel sulfate van, nickel sulfate niso4
PubChem CID 24586
ChEBI CHEBI:53001
IUPAC Name nickel(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Color Green
pH 4.3 to 4.7
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NiSO4·xH2O (x≈6)
UN Number UN3288
Merck Index 14,6517
Solubility Information Soluble in water (29.3g/100cc water at 0°C and 83.7g/100cc water at 100°C. Insoluble in alcohol,ether & acetone.
Formula Weight 154.77 (Anhydrous)
Percent Purity 99.9985%
Grade Puratronic
Odor Odorless
Density 2.070 g/mL
Assay (metals basis)
Chemical Name or Material Nickel(II) sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.