missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Product Code. 15497378
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497378 1 g 1g
15407388 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497378 Supplier Thermo Scientific Alfa Aesar Supplier No. H60878.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

It is used as a pharmaceutical intermediate and as OLED chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and as OLED chemicals.

Solubility
Partly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60871-84-3
Molecular Formula C2F6NiO6S2
Molecular Weight (g/mol) 356.82
MDL Number MFCD00673740
InChI Key KVRSDIJOUNNFMZ-UHFFFAOYSA-L
Synonym nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate
PubChem CID 9820095
IUPAC Name nickel(2+);trifluoromethanesulfonate
SMILES [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 100°C to 106°C
Linear Formula Ni(OSO2CF3)2
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Partly soluble in water.
Formula Weight 356.83
Percent Purity 96%
Chemical Name or Material Nickel(II) trifluoromethanesulfonate
compliance-icons compliance-icons
Nombre del producto
  • Nickel(II) trifluoromethanesulfonate
Palabra de advertencia
  • Danger
Clasificación
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: Nickel(II) trifluoromethanesulfonate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.