missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Metal Sheet, Pure, about 0.5mm, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
Specifications
Specifications
| Melting Point | 1455°C |
| Boiling Point | 2732°C |
| Quantity | 250 g |
| Packaging | Plastic container |
| Vapor Pressure | 1mmHg 1810 |
| Form | Solid |
| Formula Weight | 58.7g/mol |
Safety and Handling
Product Identifier
- Nickel, spheres, 5 to 11 mm
- Danger
- Carcinogenicity Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- H351-Suspected of causing cancer.
- H317-May cause an allergic skin reaction.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction