missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo/N, AL-6XN, N08367, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo/N, AL-6XN, N08367, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Tin/Titanium/Carbon/Phosphorus/Aluminium/Tungsten/Vanadium/Cobalt/Niobium/Copper/Nitrogen/Silicon/Manganese/Molybdenum/Chromium/Nickel/Iron/Beryllium/Cerium/Indium/Palladium/Ruthenium/Selenium/Magnesium/Silver/Sodium/Yttrium/Lead/Tellurium/Arsenic/Calcium/Cadmium/Bismuth/Hafnium/Hydrogen/Lanthanum/Zinc/Rhenium/Antimony/Germanium/Zirconium/Gallium/Tantalum/Oxygen |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0007 %/Tin 0.0036 %/Titanium 0.009 %/Carbon 0.0154 %/Phosphorus 0.016 %/Aluminium 0.02 %/Tungsten 0.036 %/Vanadium 0.05 %/Cobalt 0.102 %/Niobium 0.149 %/Copper 0.196 %/Nitrogen 0.203 %/Silicon 0.28 %/Manganese 0.626 %/Molybdenum 6.08 %/Chromium 20.31 %/Nickel 23.9 %/Iron 48 %/Beryllium (<0.0001) %/Cerium (<0.0001) %/Indium (<0.0001) %/Palladium (<0.0001) %/Ruthenium (<0.0001) %/Selenium (<0.0002) %/Magnesium (<0.0003) %/Silver (<0.0005) %/Sodium (<0.0005) %/Yttrium (<0.001) %/Lead (<0.002) %/Tellurium (<0.002) %/Arsenic (<0.004) %/Calcium (<0.005) %/Cadmium (<0.02) %/Bismuth (<0.03) %/Hafnium (<0.07) %/Hydrogen (<0.07) %/Lanthanum (<0.07) %/Zinc (0.0001) %/Rhenium (0.0005) %/Antimony (0.0007) %/Germanium (0.001) %/Zirconium (0.001) %/Gallium (0.003) %/Tantalum (0.003) %/Oxygen (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction