missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo/W, Alloy 686, N06686, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo/W, Alloy 686, N06686, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Oxygen/Phosphorus/Nitrogen/Carbon/Magnesium/Cobalt/Vanadium/Niobium/Silicon/Titanium/Iron/Manganese/Aluminium/Tungsten/Molybdenum/Chromium/Nickel/Silver/Yttrium/Cadmium/Bismuth/Calcium/Zinc/Tin/Lead/Selenium/Arsenic/Hafnium/Antimony/Boron/Copper/Zirconium/Tantalum |
| CAS | 7440-02-0 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.0006 %/Phosphorus 0.005 %/Nitrogen 0.006 %/Carbon 0.0079 %/Magnesium 0.008 %/Cobalt 0.01 %/Vanadium 0.01 %/Niobium 0.015 %/Silicon 0.04 %/Titanium 0.052 %/Iron 0.1 %/Manganese 0.229 %/Aluminium 0.32 %/Tungsten 3.9 %/Molybdenum 16.1 %/Chromium 20.4 %/Nickel 58.7 %/Silver (<0.0001) %/Yttrium (<0.0001) %/Cadmium (<0.0005) %/Bismuth (<0.001) %/Calcium (<0.001) %/Zinc (<0.001) %/Tin (<0.003) %/Lead (<0.004) %/Selenium (<0.005) %/Arsenic (<0.01) %/Hafnium (<0.01) %/Antimony (<0.013) %/Boron (0.003) %/Copper (0.003) %/Zirconium (0.004) %/Tantalum (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction