Learn More
Nickel, Ni/Mo, Alloy B-2, N10665, ARMI∣MBH
Product Code. 30324475
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
30324475 1 Ea. Each
1 options
Product Code. 30324475 Supplier ARMI/MBH Supplier No. IARMNIB223X
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Nickel, Ni/Mo, Alloy B-2, N10665
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aluminium, Boron, Carbon, Chromium, Cobalt, Copper, Hydrogen, Iron, Magnesium, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Phosphorus, Silicon, Sulfur, Tantalum, Tin, Titanium, Tungsten, Vanadium, Zirconium
Quantity 1 Ea.
Diameter (Metric) Outside 38 mm
Form Disc
Thickness (Metric) 3 mm
Concentration or Composition (by Analyte or Components) Silicon: 667 ppm, Vanadium: 50 ppm, Copper: 38 ppm, Phosphorus: 36 ppm, Tantalum: 25 ppm, Zirconium: 15 ppm, Niobium: 14 ppm, Oxygen: 13 ppm, Tin: 9 ppm, Hydrogen: 2 ppm, Nitrogen: 0.0007%, Sulfur: 0.0007%, Magnesium: 0.0012%, Boron: 0.0018%, Carbon: 0.003%, Tungsten: 0.0042%, Titanium: 0.021%, Cobalt: 0.116%, Aluminium: 0.13%, Manganese: 0.504%, Chromium: 0.579%, Iron: 1.36%, Molybdenum: 27.1%, Nickel: 70.2%