missing translation for 'onlineSavingsMsg'
Learn More

Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis)

Product Code. 11320048
Change view
Click to view available options
Quantity:
50 mm
150 mm
Unit Size:
150mm
50mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11320048 50 mm 50mm
11330048 150 mm 150mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320048 Supplier Thermo Scientific Alfa Aesar Supplier No. 042835.DJ
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 50 mm
Odor Odorless
Weight ≈1.75g/cm
Form Rod
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Nickel rod, 5mm (0.2 in.) dia.
What is the weight of the 150 mm size Nickel rod, 5mm (0.2in) dia, Puratronic, 99.999% (metals basis)?

The weight of the 150 mm nickel rod is ≈26.25 g.

What is the weight of the 50 mm size Nickel rod, 5mm (0.2in) dia, Puratronic, 99.999% (metals basis)?

The weight of the 50 mm nickel rod is ≈8.75 g.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.