missing translation for 'onlineSavingsMsg'
Learn More

Nickelous Carbonate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15518054 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Product Code. Quantity Packaging unitSize
15518054 500 g Poly Bottle 500g
1 options
This item is not returnable. View return policy

Product Code. 15518054

Brand: Avantor J.T.Baker 276401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 3333-67-3
Physical Form Powder
Quantity 500 g
Molecular Formula CNiO3
MDL Number MFCD00011144
Synonym nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3
InChI Key ZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES [Ni++].[O-]C([O-])=O
IUPAC Name nickel(2+);carbonate
Molecular Weight (g/mol) 118.70
PubChem CID 18746
Formula Weight 118.7
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Chemical Name or Material Nickelous Carbonate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.