missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Niclosamide

Product Code. 18064982
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18064982 Supplier R&D Systems™ Supplier No. 4079/50
  • ONLINE EXCLUSIVE PRICE
    138.12 €
    123.90 € / 50mg
    Save 14.22 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. STAT3 inhibitor; also inhibits mTORC1 signaling

Inhibitor of the STAT3 signaling pathway; inhibits the activation, nuclear translocation and transactivation of STAT3. Displays selectivity for STAT3 over STAT1, STAT5, JAK1, JAK2 and Src kinases. Inhibits the transcription of STAT3 target genes and induces cell growth inhibition, apoptosis and cell cycle arrest of cancer cells with constitutively active STAT3. Also reversibly inhibits mTORC1 signaling and stimulates autophagy in vitro; displays antineoplastic effects in acute myelogenous leukemia (AML) stem cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-65-7
Quantity 50 mg
Product Line Tocris
Molecular Formula C13H8Cl2N2O4
Formula Weight 325.99
Purity >99%
Molecular Weight (g/mol) 327.12
Chemical Name or Material Niclosamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.