missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Nicotinamide

Product Code. 18716225
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18716225 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18716225 Supplier R&D Systems™ Supplier No. TB4106RMU/500
  • ONLINE EXCLUSIVE PRICE
    509.50 €
    460.95 € / 500mg
    Save 48.55 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Nicotinamide synthesized to Ancillary Material Grade

TB4106-RMU is an Ancillary Material Grade version of Nicotinamide (Cat. No. 4106). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Find out more about our Ancillary Material Grade product range.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridine-3-carboxamide
CAS 98-92-0
Quantity 500 mg
Product Line Tocris
Target PARP
Molecular Formula C6H6N2O
Purity 0.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.