missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Nicotinamide

Product Code. 17100012
Encompass_Preferred
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 17100012

Brand: STEMCELL Technologies 07154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For pancreatic differentiation of mouse ES and iPS cells

  • Nicotinamide (NAM) is a cell culture supplement used in the differentiation of embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • It is an amide derivative of vitamin B3, a poly (ADP-ribose) polymerase (PARP) inhibitor, and represents the primary precursor of NAD+
  • Nicotinamide has also been shown to modulate stem cell differentiation in various applications, most notably for the differentiation of pancreatic mouse ES and iPS cells to pancreatic islet-like insulin-secreting cells

Specifications

Cell Type Endoderm, PSC-Derived, Pancreatic Cells, Pluripotent Stem Cells
For Use With (Application) Differentiation, Expansion
Quantity 100 g
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.