missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide Adenine Dinucleotide, MP Biomedicals™

Product Code. 11497550 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
10 g
Unit Size:
100mg
10g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497550 500 mg 500mg
11487550 100 mg 100mg
11417560 5 g 5g
11427560 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11497550 Supplier MP Biomedicals Supplier No. 02100499.5
  • ONLINE EXCLUSIVE PRICE
    159.00 €
    150.15 € / 500mg
    Save 8.85 €
    5% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acts as a major electron acceptor molecule in biological oxidations. β-Nicotinamide Adenine Dinucleotide, MP Biomedicals serves as a coenzyme of the dehydrogenases and acts as a hydrogen acceptor. β-NAD is one of the biologically active forms of

β-NAD occurs in living cells primarily in the oxidized state.

  • Serves as a coenzyme of hydride-transfer enzymes in dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • Usually acts as a hydrogen acceptor, forming NADH, which then serves as a hydrogen donor in the respiratory chain
  • Used as a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins
  • Recently reported to be a P2Y11 agonist

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Absorbance λ max (water): 259 ± 3nm
Melting Point 160°C (literature)
pH 2 to 4 (1% aq. soln.)
Quantity 500 mg
Solubility Information Soluble in water (200mg/mL)
Formula Weight 663.4 (anhydrous)
Percent Purity ≥98%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.