missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide adenine dinucleotide, oxidized Form, free acid, >98%, MP Biomedicals™

Product Code. 11407560 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407560 Supplier MP Biomedicals Supplier No. 0210049901
  • ONLINE EXCLUSIVE PRICE
    262.00 €
    247.80 € / 1g
    Save 14.20 €
    5% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 53-84-9
Color White to Yellow
Assay Percent Range >98%
Molecular Formula C21H27N7O14P2
MDL Number MFCD00150377
Quantity 1 g
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 663.43
PubChem CID 15938971
ChEBI CHEBI:57540
Formula Weight 663.4 (Anhydrous)
Physical Form Lyophilized Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide, oxidized form, free acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.