missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate

Product Code. 15845348 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
250mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15845348 500 mg Plastic bottle 500mg
15835348 250 mg Plastic bottle 250mg
15815198 1 g Plastic bottle 1g
15825198 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15845348 Supplier Thermo Scientific Supplier No. J22655.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Denna artikel kan inte returneras. Se returpolicy

CAS: 1184-16-3 | C21H27N7NaO17P3 | 765.39 g/mol

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1184-16-3
Molecular Formula C21H27N7NaO17P3
Molecular Weight (g/mol) 765.39
MDL Number MFCD00036973
InChI Key JNUMDLCHLVUHFS-QYZPTAICSA-M
Synonym nadp sodium salt, beta-nicotinamide adenine dinucleotide phosphate sodium
PubChem CID 131845262
IUPAC Name [[(2 R,3 R,4 R,5 R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2 R,3 S,4 R,5 R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodiμm
SMILES [Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)(O)=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifikationer

Physical Form Powder
Quantity 500 mg
Beilstein 4779954
Formula Weight 837.45
Packaging Plastic bottle
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt tetrahydrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.