missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%, MP Biomedicals™

Product Code. 11472492 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
1 g
Unit Size:
1g
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472492 50 mg 50mg
11462492 25 mg 25mg
11412502 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472492 Supplier MP Biomedicals Supplier No. 0215174250
  • ONLINE EXCLUSIVE PRICE
    199.00 €
    187.95 € / 50mg
    Save 11.05 €
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2646-71-1
Molecular Formula C21H26N7Na4O17P3
Molecular Weight (g/mol) 833.35
MDL Number MFCD10567216
InChI Key WYWWVJHQDVCHKF-NRTBITFTNA-J
Synonym nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form
PubChem CID 131673986
SMILES [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 50 mg
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Formula Weight 833.4
Percent Purity 0.95
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.