missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide adenine dinucleotide, reduced, disodium salt, >99%, MP Biomedicals™

Product Code. 11308789 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11308789 Supplier MP Biomedicals Supplier No. 0210116801
  • ONLINE EXCLUSIVE PRICE
    254.00 €
    240.45 € / 1g
    Save 13.55 €
    5% Off
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 606-68-8
Color White to Yellow
Assay Percent Range >99%
Molecular Formula C21H27N7Na2O14P2
MDL Number MFCD00036200
Quantity 1 g
Synonym beta-nadh disodium salt
InChI Key QRGNQKGQENGQSE-WUEGHLCSSA-L
SMILES [Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Molecular Weight (g/mol) 709.41
PubChem CID 131673989
Formula Weight 709.4
Physical Form Powder
Chemical Name or Material β-Nicotinamide adenine dinucleotide, reduced, disodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.