missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-oxide, MedChemExpress

Product Code. 16434596
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434596 Supplier Medchem Express Supplier No. HY101407
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nicotinamide N-oxide, an in vivo nicotinamide metabolite, is a potent, and selective antagonist of the CXCR2 receptor.

Nicotinamide N-oxide, an in vivo nicotinamide metabolite, is a potent, and selective antagonist of the CXCR2 receptor.

Specifications

Chemical Name or Material Nicotinamide N-oxide
CAS 1986-81-8
Color White
Purity Grade Notes Research
Molecular Formula C6H6N2O2
Quantity 100 mg
Solubility Information H2O : 20 mg/mL (144.80 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (72.40 mM; Need ultrasonic)
SMILES O=C(C1=C[N+]([O-])=CC=C1)N
Molecular Weight (g/mol) 138.12
Formula Weight 138.12
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.