missing translation for 'onlineSavingsMsg'
Learn More

Nicotine Benzoate, TRC

Product Code. 30424119
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424119 100 mg 100mg
30409208 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424119 Supplier Toronto Research Chemicals Supplier No. TRCN997715100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotine Benzoate
CAS 88660-53-1
Molecular Formula C10 H14 N2 . C7 H6 O2
Quantity 100 mg
Synonym Nicotine Benzoate, Benzoic acid','compd. with (S)-3-(1-methyl-2-pyrrolidinyl)pyridine (1:1), Pyridine','3-(1-methyl-2-pyrrolidinyl)-','(S)-','monobenzoate (9CI)
InChI Formula InChI=1S/C10H14N2.C7H6O2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;8-7(9)6-4-2-1-3-5-6/h2,4,6,8,10H,3,5,7H2,1H3;1-5H,(H,8,9)/t10-;/m0./s1
SMILES CN1CCC[C@H]1c2cccnc2.OC(=O)c3ccccc3
IUPAC Name benzoic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular Weight (g/mol) 284.353
Formula Weight 284.152
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.