missing translation for 'onlineSavingsMsg'
Learn More

Nicotine ditartrate dihydrate, 98%

Product Code. 10641853 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10641853

Brand: Thermo Scientific Acros 415660500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6019-06-3 | C18H30N2O14 | 498.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-3-6019
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
Synonym (-)-Nicotine di-(+)-hydrogen tartrate dihydrate
IUPAC Name N-{3-[(2Z)-5-chloro-2,3-dihydro-1,3-benzoxazol-2-ylidene]-4-oxocyclohexa-1,5-dien-1-yl}-2-methylpropanamide
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 97.0°C to 100.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C18H30N2O14
Quantity 50 g
Solubility Information Solubility in water: 50g/L (20°C)
Formula Weight 498.44
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nicotine ditartrate dihydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nicotine ditartrate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.