missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid hydrazide, 97%

Product Code. 10583881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10583881 25 g Plastic bottle 25g
10437540 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10583881 Supplier Thermo Scientific Acros Supplier No. 148200250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 553-53-7 | C6H7N3O | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-53-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006383
InChI Key KFUSANSHCADHNJ-UHFFFAOYSA-N
Synonym nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
PubChem CID 11112
IUPAC Name pyridine-3-carbohydrazide
SMILES NNC(=O)C1=CC=CN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160°C to 163°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 25 g
Beilstein 22, 41
Merck Index 09, 6345
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene
Formula Weight 137.14
Percent Purity 97%
Physical Form Fine Needle-like Crystalline Powder
Chemical Name or Material Nicotinic acid hydrazide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nicotinic acid hydrazide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.