missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, 98%

Product Code. 15495109
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495109 5 g 5g
15405119 25 g 25g
15415119 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495109 Supplier Thermo Scientific Alfa Aesar Supplier No. J62811.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-type calcium channel blocker; induces apoptosis in human glioblastoma cells | CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol

L-type calcium channel blocker; induces apoptosis in human glioblastoma cells.Nifedipine is a dihydropyridine calcium channel blocker used as L-type calcium channel blocker, antianginal and an antihypertensive . It induces apoptosis in human glioblastoma cells. It is used as a coronary vasodilator in the treatment of cardiac and circulatory disorders. Further, it is used in high-altitude medicine to treat high altitude pulmonary edema, hypertension and chronic stable angina. In addition, it is frequently used as a tocolytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-type calcium channel blocker; induces apoptosis in human glioblastoma cells.Nifedipine is a dihydropyridine calcium channel blocker used as L-type calcium channel blocker, antianginal and an antihypertensive . It induces apoptosis in human glioblastoma cells. It is used as a coronary vasodilator in the treatment of cardiac and circulatory disorders. Further, it is used in high-altitude medicine to treat high altitude pulmonary edema, hypertension and chronic stable angina. In addition, it is frequently used as a tocolytic agent.

Solubility
Soluble in acetone, methylene chloride, chloroform, ethyl acetate, dimethyl sulfoxide and methanol. Sparingly soluble in ethanol. Insoluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, caustics, aliphatic amines, amides, diethylamine, potassium hydride and potassium hydroxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17H18N2O6
Molecular Weight (g/mol) 346.34
MDL Number MFCD00057326
InChI Key HYIMSNHJOBLJNT-UHFFFAOYSA-N
Synonym nifedipine, procardia, adalat, procardia xl, adalat cc, cordipin, corinfar, fenihidine, citilat, oxcord
PubChem CID 4485
ChEBI CHEBI:7565
IUPAC Name 3,5-dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OC

Specifications

Melting Point 171°C to 175°C
Color White to Yellow
Quantity 5 g
Merck Index 14,6528
Solubility Information Soluble in acetone,methylene chloride,chloroform,ethyl acetate,dimethyl sulfoxide and methanol. Sparingly soluble in ethanol. Insoluble in water.
Formula Weight 346.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nifedipine
compliance-icons
Product Identifier
  • Nifedipine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Nifedipine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.