missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, TRC

Product Code. 17911695
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911695 50 mg 50mg
17942025 100 mg 100mg
17935075 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17911695 Supplier Toronto Research Chemicals Supplier No. TRCN45700050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21829-25-4
Molecular Formula C17 H18 N2 O6
Molecular Weight (g/mol) 346.33
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-, dimethyl ester (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-, dimethyl ester (8CI), 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester, 2,6-Dimethyl-3,5-dicarbomethoxy-4-(2-nitrophenyl)-1,4-dihydropyridine, 2,6-Dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylic acid dimethyl ester, 4-(2-Nitrophenyl)-2,6-dimethyl-3,5-dicarbomethoxy-1,4-dihydropyridine, Adalat, Adalat 10, Adalat 20, Adalat 5, Adalat CC, Adalat CR, Adalat Crono, Adalat FT, Adalat GITS, Adalat GITS 30, Adalat LA, Adalat LP, Adalat Oros, Adalat PA, Adalat retard, Adalate, Adapine, Adapress, Alat, Aldipin, Alfadat, Alonix, Alonix S, Alpha-Nifedipine Retard, Angipec, Anifed, Anpine, Apo-Nifed, Aprical, BAY 1040, BAY-a 1040, Bonacid, Calbloc retard, Calcibloc, Calcigard, Calcigard retard, Calciguard, Calcilat, Camont, Cardifen, Cardilat, Cardilate, Cardionorm, Cardules retard, Chronadalate, Chronadalate LP, Citilat, Coracten, Coral, Cordafen, Cordaflex, Cordalat, Cordicant, Cordilan, Cordipin, Cordipin-retard, Corinfar, Corinfar retard, Corotrend, Corynphar, Depicor SR, Depin, Depin retard, Dignokonstant, Dilafed, Dilcor, Dimethyl 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Dimethyl 4-(o-Nitrophenyl)-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, Dipinkor, Duranifin, Ecodipi, Ecodipin, Ecodipin E, Fedcor, Fedcor Retard, Fedipin, Fenamon, Fenamon SR, Fenihidine, Glopir, Hadipine, Hexadilat, Introcar, Kordafen, Macorel, Megalat, Myogard, NIfedilat, Nedipin, Nicardia, Nicardia retard, Nifangin, Nifar, Nifdemin, Nifebene, Nifecard, Nifecor, Nifedate, Nifedepat, Nifedicor, Nifedin, Nifedine, Nifedine SR, Nifedipine, Nifedipine retard, Nifedipres, Nifedirex LP, Nifelan, Nifelantern, Nifelat, Nifelat Q, Nifelate, Nifelet retard, Nifensar XL, Nificard, Nifidine, Nifipen, Niphedipine, Normadil, Novo Nifedin, Nyefax, Nyefax Retard, Orix, Osmo-Adalat, Oxcord, Pidilat, Procardia, Procardia XL, Sepamit, Sepamit R, Slofedipine XL, TN 873R, Tensopin, Tibricol, UV-ENP 349PINA, Vasdalat, Vasdalat Retard, Vasofed, Xinran, Zenusin
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)OC)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3
Formula Weight 346.1165
Percent Purity >95%
Chemical Name or Material Nifedipine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.